accessibility menu, dialog, popup

UserName

CAS RN 13826-93-2

KKBrHPdBrHBrHHBr

CAS RN 13826-93-2

IUPAC Name: dipotassium tetrabromopalladiumdiuide
Synonyms: dipotassium tetrabromopalladiumdiuide
Molecular Weight (g/mol): 508.26
Molecular Formula: Br4H4K2Pd
InChi Key: PBPKCWAOLDCQKM-UHFFFAOYSA-N
SMILES: [K+].[K+].Br.Br.Br.Br.[Pd--]
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results