accessibility menu, dialog, popup

UserName

CAS RN 13820-53-6

NaNaClPdClClCl

CAS RN 13820-53-6

IUPAC Name: platinum(2+) disodium tetrachloride
Synonyms: sodium tetrachloropalladate
Molecular Weight (g/mol): 382.86
Molecular Formula: Cl4Na2Pt
InChi Key: UXSGJRDAUMCHRP-UHFFFAOYSA-J
SMILES: [Na+].[Na+].[Cl-].[Cl-].[Cl-].[Cl-].[Pt++]
Molecular Weight (g/mol) 
  • (8)
  • (1)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
Percent Purity 
  • (3)
  • (3)
  • (2)
Physical Form 
  • (5)
  • (3)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (5)
  • (1)
  • (8)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (3)
  • (3)
  • (2)
  • (5)
  • (3)
  • (2)