accessibility menu, dialog, popup

UserName

PtClClClClNH4NH4

CAS RN 13820-41-2

IUPAC Name: —
Synonyms: —
Molecular Weight (g/mol): 336.88
Molecular Formula: Cl4Pt
InChi Key: INXDHMXCTAMKSX-UHFFFAOYSA-N
SMILES: [Cl].[Cl].[Cl].[Cl].[Pt++]
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results