CAS RN 1378757-22-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Alpha-(Chloromethyl)-3-hydroxybenzenemethanol, TRC
CAS: 1378757-22-2 Molecular Formula: C8H9ClO2 Molecular Weight (g/mol): 172.61 Synonym: α-(Chloromethyl)-3-hydroxybenzenemethanol,(RS)-2-Chloro-1-(3-hydroxyphenyl)ethanol IUPAC Name: 3-(2-chloro-1-hydroxyethyl)phenol SMILES: OC(CCl)c1cccc(O)c1