accessibility menu, dialog, popup

UserName

NS

CAS RN 13781-53-8

IUPAC Name: 2-(thiophen-3-yl)acetonitrile
Synonyms: 3-cyanomethylthiophene
Molecular Weight (g/mol): 123.17
Molecular Formula: C6H5NS
InChi Key: GWZCLMWEJWPFFA-UHFFFAOYSA-N
SMILES: N#CCC1=CSC=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results