accessibility menu, dialog, popup

UserName

H2OH2OH2OH2OH2OH2OH2OH2OOSOOOOSOOOOSOOOTmTm

CAS RN 13778-40-0

IUPAC Name: dithulium(3+) octahydrate trisulfate
Synonyms: dithulium(3+) octahydrate trisulfate
Molecular Weight (g/mol): 770.16
Molecular Formula: H16O20S3Tm2
InChi Key: GPBYLGMJMIXKMU-UHFFFAOYSA-H
SMILES: O.O.O.O.O.O.O.O.[Tm+3].[Tm+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)
Grade 
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)