accessibility menu, dialog, popup

UserName

OOF

CAS RN 137736-06-2

IUPAC Name: 4-(4-fluorophenoxy)benzaldehyde
Synonyms: 4-(4-fluorophenoxy)benzaldehyde
Molecular Weight (g/mol): 216.21
Molecular Formula: C13H9FO2
InChi Key: YUPBWHURNLRZQL-UHFFFAOYSA-N
SMILES: FC1=CC=C(OC2=CC=C(C=O)C=C2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results