accessibility menu, dialog, popup

UserName

BaOPOOOPOO

CAS RN 13762-83-9

IUPAC Name: —
Synonyms: —
Molecular Weight (g/mol): 295.27
Molecular Formula: BaO6P2
InChi Key: RYSCSJQMVDRYDV-UHFFFAOYSA-J
SMILES: [Ba+4].[O-][P]([O-])=O.[O-][P]([O-])=O
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results