accessibility menu, dialog, popup

UserName

BHHHHK

CAS RN 13762-51-1

IUPAC Name: potassium λ¹-boranuidylidene
Synonyms: potassium λ¹-boranuidylidene
Molecular Weight (g/mol): 49.91
Molecular Formula: BK
InChi Key: HHUJLKPGQMFFMS-UHFFFAOYSA-N
SMILES: [B-].[K+]
Quantity 
  • (1)
  • (3)
  • (2)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
Physical Form 
  • (4)
  • (3)
  • (4)
Percent Purity 
  • (4)
  • (3)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (4)
  • (7)
  • (3)
  • (1)
  • (3)
  • (2)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (4)
  • (3)
  • (4)
  • (4)
  • (3)
  • (3)