accessibility menu, dialog, popup

UserName

CAS RN 13759-83-6

IUPAC Name: samarium(3+) hexahydrate trinitrate
Synonyms: samarium(3+) hexahydrate trinitrate
Molecular Weight (g/mol): 444.46
Molecular Formula: H12N3O15Sm
InChi Key: HDCOFJGRHQAIPE-UHFFFAOYSA-N
SMILES: O.O.O.O.O.O.[Sm+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
Physical Form 
  • (2)
  • (3)
  • (2)
Quantity 
  • (3)
  • (3)
  • (1)
Grade 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (5)
  • (2)
  • (3)
  • (2)
  • (3)
  • (3)
  • (1)
  • (3)
  • (2)
  • (2)
  • (3)