accessibility menu, dialog, popup

UserName

NFFCl

CAS RN 1374659-44-5

IUPAC Name: 3-chloro-4-(difluoromethyl)pyridine
Synonyms: 3-chloro-4-(difluoromethyl)pyridine
Molecular Weight (g/mol): 163.55
Molecular Formula: C6H4ClF2N
InChi Key: UZRBGKNPGFNLDR-UHFFFAOYSA-N
SMILES: FC(F)C1=C(Cl)C=NC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results