accessibility menu, dialog, popup

UserName

H3COFFCl

CAS RN 1373921-04-0

IUPAC Name: 1-chloro-3,4-difluoro-2-methoxybenzene
Synonyms: 1-chloro-3,4-difluoro-2-methoxybenzene
Molecular Weight (g/mol): 178.56
Molecular Formula: C7H5ClF2O
InChi Key: ZLXLBDWCQIFFMA-UHFFFAOYSA-N
SMILES: COC1=C(Cl)C=CC(F)=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results