accessibility menu, dialog, popup

UserName

CH3NFFFF

CAS RN 1373920-75-2

IUPAC Name: 2-[4-fluoro-3-methyl-5-(trifluoromethyl)phenyl]acetonitrile
Synonyms: 2-[4-fluoro-3-methyl-5-(trifluoromethyl)phenyl]acetonitrile
Molecular Weight (g/mol): 217.17
Molecular Formula: C10H7F4N
InChi Key: YKXAXCYASRRQID-UHFFFAOYSA-N
SMILES: CC1=C(F)C(=CC(CC#N)=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results