accessibility menu, dialog, popup

UserName

CAS RN 1373920-67-2

CH3OOFFFF

CAS RN 1373920-67-2

IUPAC Name: 2-fluoro-5-methoxy-3-(trifluoromethyl)benzaldehyde
Synonyms: 2-fluoro-5-methoxy-3-(trifluoromethyl)benzaldehyde
Molecular Weight (g/mol): 222.14
Molecular Formula: C9H6F4O2
InChi Key: YHTRIRREEHYBRI-UHFFFAOYSA-N
SMILES: COC1=CC(=C(F)C(C=O)=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results