accessibility menu, dialog, popup

UserName

CH3ONFFFF

CAS RN 1373920-64-9

IUPAC Name: 2-[2-fluoro-5-methoxy-3-(trifluoromethyl)phenyl]acetonitrile
Synonyms: 2-[2-fluoro-5-methoxy-3-(trifluoromethyl)phenyl]acetonitrile
Molecular Weight (g/mol): 233.17
Molecular Formula: C10H7F4NO
InChi Key: PRSLVVFXWPHVSP-UHFFFAOYSA-N
SMILES: COC1=CC(=C(F)C(CC#N)=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results