accessibility menu, dialog, popup

UserName

CAS RN 13720-06-4

IUPAC Name: 2,6-dibromonaphthalene
Synonyms: 2,6-dibromonaphthalene
Molecular Weight (g/mol): 285.97
Molecular Formula: C10H6Br2
InChi Key: PJZDEYKZSZWFPX-UHFFFAOYSA-N
SMILES: BrC1=CC2=CC=C(Br)C=C2C=C1
Quantity 
  • (2)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results