accessibility menu, dialog, popup

UserName

H3CH3CCH3PCH3CH3H3CCH3CH3H3C

CAS RN 13716-12-6

IUPAC Name: tri-tert-butylphosphane
Synonyms: tri-tert-butylphosphane
Molecular Weight (g/mol): 202.32
Molecular Formula: C12H27P
InChi Key: BWHDROKFUHTORW-UHFFFAOYSA-N
SMILES: CC(C)(C)P(C(C)(C)C)C(C)(C)C
Boiling Point 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)