CAS RN 137049-02-6
CAS RN 137049-02-6
IUPAC Name:
1,2-dimethyl-1H-imidazole-4-sulfonyl chloride
Synonyms:
1,2-dimethylimidazole-4-sulfonyl chloride
Molecular Weight (g/mol):
194.63
Molecular Formula:
C5H7ClN2O2S
InChi Key:
JPAFTHVNSBWVQJ-UHFFFAOYSA-N
SMILES:
CN1C=C(N=C1C)S(Cl)(=O)=O
1
–
3
of
3
results
1
1,2-Dimethyl-1H-imidazole-4-sulfonyl Chloride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
1,2-Dimethyl-1H-imidazole-4-sulfonyl Chloride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
1,2-Dimethyl-1H-imidazole-4-sulfonyl Chloride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.