missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dimethyl-1H-imidazole-4-sulfonyl Chloride, TRC

Product Code. 30375121
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30375121 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30375121 Supplier Toronto Research Chemicals Supplier No. TRCD47416325MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1,2-Dimethyl-1H-imidazole-4-sulfonyl Chloride
CAS 137049-02-6
Molecular Formula C5 H7 Cl N2 O2 S
Quantity 25 mg
Synonym 1,2-Dimethylimidazole-4-Sulfonyl Chloride
InChI Formula InChI=1S/C5H7ClN2O2S/c1-4-7-5(3-8(4)2)11(6,9)10/h3H,1-2H3
SMILES Cc1nc(cn1C)S(=O)(=O)Cl
IUPAC Name 1,2-dimethylimidazole-4-sulfonyl chloride
Molecular Weight (g/mol) 194.639
Formula Weight 193.992
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.