accessibility menu, dialog, popup

UserName

CAS RN 137-07-5

NH2HS

CAS RN 137-07-5

IUPAC Name: hydrogen 2-aminobenzene-1-thiol chloride
Synonyms: 2-aminothiophenol
Molecular Weight (g/mol): 161.65
Molecular Formula: C6H8ClNS
InChi Key: PEDOSWZBWWCOAY-UHFFFAOYSA-N
SMILES: [H+].[Cl-].NC1=CC=CC=C1S
Quantity 
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (3)
  • (2)
  • (2)