accessibility menu, dialog, popup

UserName

CH3SO

CAS RN 13679-70-4

IUPAC Name: 5-methylthiophene-2-carbaldehyde
Synonyms: 5-methylthiophene-2-carbaldehyde
Molecular Weight (g/mol): 126.17
Molecular Formula: C6H6OS
InChi Key: VAUMDUIUEPIGHM-UHFFFAOYSA-N
SMILES: CC1=CC=C(S1)C=O
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)