accessibility menu, dialog, popup

UserName

CAS RN 13671-00-6

CH3OOFF

CAS RN 13671-00-6

IUPAC Name: methyl 2,6-difluorobenzoate
Synonyms: methyl 2,6-difluorobenzoate
Molecular Weight (g/mol): 172.13
Molecular Formula: C8H6F2O2
InChi Key: QNPFLTKQLFSKBY-UHFFFAOYSA-N
SMILES: COC(=O)C1=C(F)C=CC=C1F
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results