accessibility menu, dialog, popup

UserName

CAS RN 1363404-84-5

IUPAC Name: ethyl 8-(trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylate
Synonyms: ethyl 8-(trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylate
Molecular Weight (g/mol): 258.2
Molecular Formula: C11H9F3N2O2
InChi Key: WZLPRVSGJRYTIQ-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CN2C=CC=C(C2=N1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results