accessibility menu, dialog, popup

UserName

CAS RN 13631-21-5

OHClBr

CAS RN 13631-21-5

IUPAC Name: 4-bromo-3-chlorophenol
Synonyms: 4-bromo-3-chlorophenol
Molecular Weight (g/mol): 207.45
Molecular Formula: C6H4BrClO
InChi Key: FQEYHIPPYOSPLF-UHFFFAOYSA-N
SMILES: OC1=CC(Cl)=C(Br)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results