CAS RN 135575-42-7
CAS RN 135575-42-7
IUPAC Name:
N-[(3S,6S,9S,11R,15S,18S,20R,21R,24S,25S)-3-[(1S)-2-carbamoyl-1-hydroxyethyl]-6-[(1S,2S)-1,2-dihydroxy-2-(4-hydroxyphenyl)ethyl]-11,20,21,25-tetrahydroxy-15-[(1S)-1-hydroxyethyl]-2,5,8,14,17,23-hexaoxo-1,4,7,13,16,22-hexaazatricyclo[22.3.0.0⁹,¹³]heptacosan-18-yl]-10,12-dimethyltetradecanamide
Synonyms:
pneumocandin bo
Molecular Weight (g/mol):
1065.23
Molecular Formula:
C50H80N8O17
InChi Key:
DQXPFAADCTZLNL-YSHLLHMASA-N
SMILES:
CCC(C)CC(C)CCCCCCCCC(=O)N[C@H]1C[C@@H](O)[C@@H](O)NC(=O)[C@@H]2[C@@H](O)CCN2C(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H]2C[C@@H](O)CN2C(=O)[C@@H](NC1=O)[C@H](C)O)[C@H](O)[C@@H](O)C1=CC=C(O)C=C1)[C@@H](O)CC(N)=O
1
–
1
of
1
results
1
Pneumocandin B0, MedChemExpress
MedChemExpress Pneumocandin B0(L-688786), a key intermediate in the synthesis of the antifungal agent, Cancidas, has led to the identification of several materials with potential for improved performance.