missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pneumocandin B0, MedChemExpress
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Description
Pneumocandin B0(L-688786), a key intermediate in the synthesis of the antifungal agent, Cancidas, has led to the identification of several materials with potential for improved performance.
Specifications
Specifications
| Chemical Name or Material | Pneumocandin B0 |
| CAS | 135575-42-7 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C50H80N8O17 |
| Quantity | 10 mg |
| Synonym | L-688786 |
| Solubility Information | DMSO : 100 mg/mL (93.88 mM; Need ultrasonic) |
| SMILES | O=C([C@](NC(C(NC([C@@](C[C@@H](O)C1)([H])N1C2=O)=O)[C@H](O)[C@H](C3=CC=C(O)C=C3)O)=O)([H])[C@H](O)CC(N)=O)N4[C@@](C(N[C@@H]([C@@H](C[C@@H](C(NC2[C@H](O)C)=O)NC(CCCCCCCC[C@@H](C)C[C@@H](C)CC)=O)O)O)=O)([H])[C@@H](O)CC4 |
| Molecular Weight (g/mol) | 1065.21 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction