CAS RN 1355729-38-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Zinc Difluoromethanesulfinate (Contains up to 1 equiv. of Zinc Chloride), TRC
CAS: 1355729-38-2 Molecular Formula: C2H2F4O4S2Zn Molecular Weight (g/mol): 295.54 Synonym: Baran Reagent IUPAC Name: zinc;difluoromethanesulfinate SMILES: [Zn+2].[O-]S(=O)C(F)F.[O-]S(=O)C(F)F