CAS RN 135531-25-8
CAS RN 135531-25-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Bromochloronitromethane (>85%), TRC
CAS: 135531-25-8 Molecular Formula: C H Br Cl N O2 Molecular Weight (g/mol): 174.38 Synonym: Bromochloronitromethane IUPAC Name: bromo-chloro-nitromethane SMILES: [O-][N+](=O)C(Cl)Br