accessibility menu, dialog, popup

UserName

CAS RN 13535-13-2

NNNH2

CAS RN 13535-13-2

IUPAC Name: 5-phenylpyrazin-2-amine
Synonyms: 5-phenylpyrazin-2-amine
Molecular Weight (g/mol): 171.2
Molecular Formula: C10H9N3
InChi Key: KJAKXVBZQBPPOB-UHFFFAOYSA-N
SMILES: NC1=NC=C(N=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results