accessibility menu, dialog, popup

UserName

CH3OFOHOCl(E/Z)

CAS RN 1353001-74-7

IUPAC Name: 3-(4-chloro-2-fluoro-3-methoxyphenyl)prop-2-enoic acid
Synonyms: 3-(4-chloro-2-fluoro-3-methoxyphenyl)prop-2-enoic acid
Molecular Weight (g/mol): 230.62
Molecular Formula: C10H8ClFO3
InChi Key: RBQIGCSSGNRGJR-UHFFFAOYSA-N
SMILES: COC1=C(Cl)C=CC(C=CC(O)=O)=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results