CAS RN 13515-65-6
CAS RN 13515-65-6
IUPAC Name:
2-methylpropanethioamide
Synonyms:
2-methylpropanethioamide
Molecular Weight (g/mol):
103.18
Molecular Formula:
C4H9NS
InChi Key:
NPCLRBQYESMUPD-UHFFFAOYSA-N
SMILES:
CC(C)C(N)=S
1
–
2
of
2
results
1
Thioisobutyramide, 95%
CAS: 13515-65-6 Molecular Formula: C4H9NS Molecular Weight (g/mol): 103.183 MDL Number: MFCD07369538 InChI Key: NPCLRBQYESMUPD-UHFFFAOYSA-N Synonym: thioisobutyramide,isobutylthioamide,isobutyrthioamide,2-methylpropaneothioamide,2-methylpropane-thioamide,2-methyl-propanethioamide,acmc-209by0,propanethioamide,2-methyl,ksc519m8d,2-methyl propanethioamide PubChem CID: 10909507 IUPAC Name: 2-methylpropanethioamide SMILES: CC(C)C(=S)N
2
2-Methylpropanethioamide, Mikromol
Discover Mikromol- your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.