accessibility menu, dialog, popup

UserName

CAS RN 135034-10-5

NNClI

CAS RN 135034-10-5

IUPAC Name: 3-chloro-6-iodopyridazine
Synonyms: 3-chloro-6-iodopyridazine
Molecular Weight (g/mol): 240.43
Molecular Formula: C4H2ClIN2
InChi Key: PNEPCDPKMXJYIQ-UHFFFAOYSA-N
SMILES: ClC1=NN=C(I)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results