accessibility menu, dialog, popup

UserName

CAS RN 13498-14-1

OPOPOClClClCl

CAS RN 13498-14-1

IUPAC Name: [(dichlorophosphoryl)oxy]phosphonoyl dichloride
Synonyms: (dichlorophosphoryl)oxyphosphonoyl dichloride
Molecular Weight (g/mol): 251.74
Molecular Formula: Cl4O3P2
InChi Key: CNTIXUGILVWVHR-UHFFFAOYSA-N
SMILES: ClP(Cl)(=O)OP(Cl)(Cl)=O
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)