CAS RN 134924-71-3
CAS RN 134924-71-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
trans-Hydroxy Praziquantel, TRC
CAS: 134924-71-3 Molecular Formula: C19H24N2O3 Molecular Weight (g/mol): 328.41 Synonym: 1,2,3,6,7,11b-Hexahydro-2-[(cis-4-hydroxycyclohexyl)carbonyl]-4H-pyrazino[2,1-a]isoquinolin-4-one,trans-1,2,3,6,7,11b-Hexahydro-2-[(4-hydroxycyclohexyl)carbonyl]- 4H-pyrazino[2,1-a]isoquinolin-4-one; IUPAC Name: 2-((1r,4r)-4-hydroxycyclohexane-1-carbonyl)-1,2,3,6,7,11b-hexahydro-4H-pyrazino[2,1-a]isoquinolin-4-one SMILES: O=C1N2CCC3=CC=CC=C3C2CN(C([C@H]4CC[C@H](O)CC4)=O)C1