accessibility menu, dialog, popup

UserName

CH3NH2ONHOO

CAS RN 13485-59-1

IUPAC Name: 1-(2-aminopropanoyl)pyrrolidine-2-carboxylic acid
Synonyms: proline,D-alanyl
Molecular Weight (g/mol): 186.21
Molecular Formula: C8H14N2O3
InChi Key: WPWUFUBLGADILS-UHFFFAOYNA-N
SMILES: CC(N)C(=O)N1CCCC1C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results