accessibility menu, dialog, popup

UserName

CAS RN 134822-76-7

IUPAC Name: 6-hydroxy-2H-chromene-3-carbaldehyde
Synonyms: 6-hydroxy-2H-chromene-3-carbaldehyde
Molecular Weight (g/mol): 176.17
Molecular Formula: C10H8O3
InChi Key: JSPGOHYLKVWWKI-UHFFFAOYSA-N
SMILES: OC1=CC2=C(OCC(C=O)=C2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results