accessibility menu, dialog, popup

UserName

H3CNHNNN

CAS RN 1347815-10-4

IUPAC Name: 2-(3-methylphenyl)-1H-imidazole-4,5-dicarbonitrile
Synonyms: 2-(3-methylphenyl)-1H-imidazole-4,5-dicarbonitrile
Molecular Weight (g/mol): 208.22
Molecular Formula: C12H8N4
InChi Key: OIQBYNHKZGZUMW-UHFFFAOYSA-N
SMILES: CC1=CC=CC(=C1)C1=NC(C#N)=C(N1)C#N
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results