accessibility menu, dialog, popup

UserName

CAS RN 1347814-85-0

H3CCH3NHNNH2HCl

CAS RN 1347814-85-0

IUPAC Name: 5-(propan-2-yl)-1H-pyrazol-3-amine hydrochloride
Synonyms: 5-isopropyl-1H-pyrazol-3-amine hydrochloride
Molecular Weight (g/mol): 161.63
Molecular Formula: C6H12ClN3
InChi Key: JHGOPXUCCFVTPD-UHFFFAOYSA-N
SMILES: Cl.CC(C)C1=CC(N)=NN1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results