accessibility menu, dialog, popup

UserName

CAS RN 13476-99-8

OOOOOOCHHCCHH3CH3CCH3H3CCH3H3CV

CAS RN 13476-99-8

IUPAC Name: 4-hydroxypent-3-en-2-one vanadium
Synonyms: 4-hydroxypent-3-en-2-one vanadium
Molecular Weight (g/mol): 151.06
Molecular Formula: C5H8O2V
InChi Key: CZFPVTPZFRXJDT-UHFFFAOYSA-N
SMILES: [V].CC(O)=CC(C)=O
Percent Purity 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results