CAS RN 1346606-59-4
CAS RN 1346606-59-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Diminazene-13C2,15N4 Dihydrochloride (major), TRC
CAS: 1346606-59-4 Molecular Formula: C1213C2H17Cl2N315N4 Molecular Weight (g/mol): 360.2 Synonym: 4,4'-(1-Triazene-1,3-diyl)bisbenzenecarboximidamide-13C2,15N4Dihydrochloride,4,4'-(Diazoamino)dibenzamidine-13C2,15N4 Dihydrochloride,Berenil-13C2,15N4 Dihydrochloride; SMILES: [15NH2][13C](C1=CC=C(C=C1)N/N=N/C2=CC=C([13C]([15NH2])=[15NH])C=C2)=[15NH]