CAS RN 13466-35-8
CAS RN 13466-35-8
IUPAC Name:
3-chloro-1,2-dihydropyridin-2-one
Synonyms:
3-chloro-1H-pyridin-2-one
Molecular Weight (g/mol):
129.54
Molecular Formula:
C5H4ClNO
InChi Key:
XTYNIPUFKBBALX-UHFFFAOYSA-N
SMILES:
ClC1=CC=CNC1=O
1
–
1
of
1
results
1
3-Chloro-2-hydroxypyridine, 97%
CAS: 13466-35-8 Molecular Formula: C5H4ClNO Molecular Weight (g/mol): 129.54 MDL Number: MFCD01646109 InChI Key: XTYNIPUFKBBALX-UHFFFAOYSA-N Synonym: 3-chloro-2-hydroxypyridine,3-chloropyridin-2-ol,3-chloro-2-pyridinol,2-hydroxy-3-chloropyridine,3-chloropyridin-2 1h-one,3-chloro-2 1h-pyridinone,chloropyridone,2 1h-pyridinone, 3-chloro,chloropyridinol,pubchem6538 PubChem CID: 582210 IUPAC Name: 3-chloro-1H-pyridin-2-one SMILES: ClC1=CC=CNC1=O