accessibility menu, dialog, popup

UserName

CAS RN 13454-84-7

CsOClOOO

CAS RN 13454-84-7

IUPAC Name: caesium(1+) perchlorate
Synonyms: caesium perchlorate
Molecular Weight (g/mol): 232.35
Molecular Formula: ClCsO4
InChi Key: WKDKOOITVYKILI-UHFFFAOYSA-M
SMILES: [Cs+].[O-][Cl](=O)(=O)=O
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results