accessibility menu, dialog, popup

UserName

CAS RN 13451-14-4

H2OH2OOHTeOOHO

CAS RN 13451-14-4

IUPAC Name: telluric acid dihydrate
Synonyms: telluric acid dihydrate
Molecular Weight (g/mol): 229.64
Molecular Formula: H6O6Te
InChi Key: VCYUGZHEVVXOPD-UHFFFAOYSA-N
SMILES: O.O.O[Te](O)(=O)=O
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results