accessibility menu, dialog, popup

UserName

CAS RN 13445-16-4

H3CONBrOCH3

CAS RN 13445-16-4

IUPAC Name: 3-bromo-2,6-dimethoxypyridine
Synonyms: 3-bromo-2,6-dimethoxypyridine
Molecular Weight (g/mol): 218.05
Molecular Formula: C7H8BrNO2
InChi Key: UUTYQBCAEWGFQL-UHFFFAOYSA-N
SMILES: COC1=NC(OC)=C(Br)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results