accessibility menu, dialog, popup

UserName

FFFPFFFFFF

CAS RN 13406-29-6

IUPAC Name: tris[4-(trifluoromethyl)phenyl]phosphane
Synonyms: tris[4-(trifluoromethyl)phenyl]phosphane
Molecular Weight (g/mol): 466.29
Molecular Formula: C21H12F9P
InChi Key: PXYCJKZSCDFXLR-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC=C(C=C1)P(C1=CC=C(C=C1)C(F)(F)F)C1=CC=C(C=C1)C(F)(F)F
Molecular Weight (g/mol) 
  • (3)
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results