accessibility menu, dialog, popup

UserName

CAS RN 13395-36-3

IUPAC Name: ethyl 5,5-dimethyl-2,4-dioxohexanoate
Synonyms: ethyl 5,5-dimethyl-2,4-dioxohexanoate
Molecular Weight (g/mol): 200.23
Molecular Formula: C10H16O4
InChi Key: NIMKIMUBJFWPTD-UHFFFAOYSA-N
SMILES: CCOC(=O)C(=O)CC(=O)C(C)(C)C
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (1)
  • (2)
  • (1)
  • (3)