accessibility menu, dialog, popup

UserName

OOCuOOH3CH3CCH3CH3

CAS RN 13395-16-9

IUPAC Name: bis(4-hydroxypent-3-en-2-one) copper
Synonyms: bis(4-hydroxypent-3-en-2-one) copper
Molecular Weight (g/mol): 263.78
Molecular Formula: C10H16CuO4
InChi Key: ZKXWKVVCCTZOLD-UHFFFAOYSA-N
SMILES: [Cu].CC(O)=CC(C)=O.CC(O)=CC(C)=O
Quantity 
  • (4)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (3)
  • (1)
  • (3)
Physical Form 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (6)
  • (3)
  • (1)
  • (4)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (3)
  • (3)
  • (3)
  • (1)