accessibility menu, dialog, popup

UserName

CAS RN 133851-52-2

H3CCH3H3COONOHO

CAS RN 133851-52-2

IUPAC Name: 1-[(tert-butoxy)carbonyl]-2,3-dihydro-1H-indole-2-carboxylic acid
Synonyms: 1-(tert-butoxycarbonyl)-2,3-dihydroindole-2-carboxylic acid
Molecular Weight (g/mol): 263.29
Molecular Formula: C14H17NO4
InChi Key: QONNUMLEACJFME-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)N1C(CC2=CC=CC=C12)C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results