accessibility menu, dialog, popup

UserName

CH3NNNH2NHN

CAS RN 1337596-50-5

IUPAC Name: 5-(2,3-dihydro-1H-indol-5-yl)-7-methyl-7H-pyrrolo[2,3-d]pyrimidin-4-amine
Synonyms: 5-(2,3-dihydro-1H-indol-5-yl)-7-methylpyrrolo[2,3-d]pyrimidin-4-amine
Molecular Weight (g/mol): 265.32
Molecular Formula: C15H15N5
InChi Key: CXRNQXFFWARLSJ-UHFFFAOYSA-N
SMILES: CN1C=C(C2=C(N)N=CN=C12)C1=CC2=C(NCC2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results