CAS RN 13373-97-2
IUPAC Name:
icosane-1-thiol
Synonyms:
icosane-1-thiol
Molecular Weight (g/mol):
314.62
Molecular Formula:
C20H42S
InChi Key:
YYHYWOPDNMFEAV-UHFFFAOYSA-N
SMILES:
CCCCCCCCCCCCCCCCCCCCS
1
–
2
of
2
results
1
1-Eicosanethiol, 98%
CAS: 13373-97-2 Molecular Formula: C20H42S Molecular Weight (g/mol): 314.616 MDL Number: MFCD07779406 InChI Key: YYHYWOPDNMFEAV-UHFFFAOYSA-N Synonym: 1-eicosanethiol,eicosanethiol,1-icosanethiol,icosyl mercaptan,eicosyl mercaptan,acmc-209btf PubChem CID: 139443 IUPAC Name: icosane-1-thiol SMILES: CCCCCCCCCCCCCCCCCCCCS
2
1-Eicosanethiol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.